Skip to Content
Merck
All Photos(1)

Key Documents

W354201

Sigma-Aldrich

6,10-Dimethyl-5,9-undecadien-2-one

≥97%, stabilized, FG

Synonym(s):

6,10-Dimethyl-5,9-undecadien-2-one, mixture of isomers, Geranylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2COCH3
CAS Number:
Molecular Weight:
194.31
FEMA Number:
3542
Beilstein:
1722277
EC Number:
Council of Europe no.:
11088
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.123
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥97%

contains

α- Tocopherol, Synthetic as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; floral; rose

SMILES string

C\C(C)=C\CC\C(C)=C\CCC(C)=O

InChI

1S/C13H22O/c1-11(2)7-5-8-12(3)9-6-10-13(4)14/h7,9H,5-6,8,10H2,1-4H3/b12-9+

InChI key

HNZUNIKWNYHEJJ-FMIVXFBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6,10-Dimethyl-5,9-undecadien-2-one is one of the key volatile flavor compounds of tomato and certain species of mushroom.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of some volatile components of seven fresh wild mushrooms (Basidiomycetes).
Rapior S, et al.
J. Essent. Oil Res., 8(2), 199-201 (1996)
Adrian Haiduc et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1152, 122228-122228 (2020-06-26)
Tobacco smoking contributes to tooth discoloration. Pigmented compounds in the smoke generated by combustion of tobacco can cause discoloration of dental hard tissues. However, aerosols from heated tobacco products cause less discoloration than cigarette smoke (CS) in vitro. The objective
Flavor of tomato and tomato products
Petro?Turza M.
Food Reviews International, 2(3), 309-351 (1986)
Volatile composition of fourteen species of fresh wild mushrooms (Boletales).
Rapior S, et al.
J. Essent. Oil Res., 9(2), 231-234 (1997)
Tomoaki Ishihara et al.
Biochemical pharmacology, 82(7), 728-736 (2011-07-09)
It is important in treatment of gastric ulcers to not only prevent further ulcer formation but also enhance ulcer healing. When cells are exposed to gastric irritants, expression of heat shock proteins (HSPs) is induced, making the cells resistant to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service