Key Documents
SYNF641425
Synthware pressure equalizing funnel with high vacuum valve
25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Synonym(s):
Synthware F614425, dropping funnel
About This Item
UNSPSC Code:
41121809
NACRES:
NB.51
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F641425
capacity
25 mL
25 mL
Related Categories
General description
With PTFE extended tip valve for semi-needle valve adjustment.
Legal Information
Synthware is a trademark of Kemtech America Inc.