774723
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H10FN · HCl
CAS Number:
Molecular Weight:
187.64
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
182-183 °C
functional group
fluoro
SMILES string
Cl.Fc1ccc2CCNCc2c1
InChI
1S/C9H10FN.ClH/c10-9-2-1-7-3-4-11-6-8(7)5-9;/h1-2,5,11H,3-4,6H2;1H
InChI key
LOJNQXIJLCPQDR-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service