692751
(R,R)-DACH-pyridyl Trost ligand
95%
Synonym(s):
(–)-N,N′-(1R,2R)-1,2-Diaminocyclohexanediylbis(2-pyridinecarboxamide)
About This Item
Recommended Products
Assay
95%
form
solid
optical activity
[α]20/D -97.0°, c = 1 in methanol
mp
171-176 °C
functional group
amide
SMILES string
O=C(N[C@@H]1CCCC[C@H]1NC(=O)c2ccccn2)c3ccccn3
InChI
1S/C18H20N4O2/c23-17(15-9-3-5-11-19-15)21-13-7-1-2-8-14(13)22-18(24)16-10-4-6-12-20-16/h3-6,9-14H,1-2,7-8H2,(H,21,23)(H,22,24)/t13-,14-/m1/s1
InChI key
BVYFDUZVVBXFBQ-ZIAGYGMSSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.
Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.
Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.
Trost group pioneers C-2 symmetric ligands for rapid chiral product synthesis in asymmetric catalytic reactions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service