All Photos(1)
About This Item
Linear Formula:
(CH3)3CC6H3(CH3)2
CAS Number:
Molecular Weight:
162.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.498 (lit.)
bp
200-210 °C (lit.)
density
0.871 g/mL at 25 °C (lit.)
SMILES string
Cc1ccc(cc1C)C(C)(C)C
InChI
1S/C12H18/c1-9-6-7-11(8-10(9)2)12(3,4)5/h6-8H,1-5H3
InChI key
QRPPSTNABSMSCS-UHFFFAOYSA-N
General description
4-tert-Butyl-o-xylene (1,2-dimethyl-4-tert-Butylbenzene, DMTBB) is a dimethylalkylbenzene. It has been synthesized by reacting o-xylene with tert-butyl alcohol over zeolite based catalytic system.
Application
4-tert-Butyl-o-xylene may be used as a starting material in the synthesis of 1,2-bis(bromomethyl )-4-tert-butylbenzene. It may also be used as a solvent in the synthesis of 2,7-di-tert-butylcarbazole, via ring closure of 4,4′-di-tert-butyl-2,2′-diaminobiphenyl in the presence of Nafion-H.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
179.6 °F - closed cup
Flash Point(C)
82 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Selective alkylation of xylenes by alcohols on zeolite catalysts.
Isakov YI, et al.
Russian Chemical Bulletin, 45(12), 2763-2768 (1996)
Poly (5-tert-butyl) benzothiophene: a soluble form of polyisothianaphthene with a large nonlinear optical response.
Drury A, et al.
Journal of Materials Chemistry, 8(11), 2353-2355 (1998)
Organic reactions catalyzed by solid superacids. 10. Perfluorinated sulfonic acid resin (Nafion-H) catalyzed ring closure reaction of 2, 2'-diaminobiphenyls. A preparative route to carbazoles.
Yamato T, et al.
The Journal of Organic Chemistry, 56(21), 6248-6250 (1991)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service