Skip to Content
Merck
All Photos(2)

Key Documents

147311

Sigma-Aldrich

2-Nitro-p-cymene

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
1-(CH3)C6H3(NO2)CH(CH3)2
CAS Number:
Molecular Weight:
179.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

liquid

impurities

7% 3-nitro-p-cymene

refractive index

n20/D 1.528 (lit.)

density

1.07 g/mL at 25 °C (lit.)

functional group

nitro

SMILES string

CC(C)c1ccc(C)c(c1)[N+]([O-])=O

InChI

1S/C10H13NO2/c1-7(2)9-5-4-8(3)10(6-9)11(12)13/h4-7H,1-3H3

InChI key

DRKFWQDBPGTSOO-UHFFFAOYSA-N

General description

Reduction products of 2-nitro-p-cymene are significant dye intermediates.

Application

2-Nitro-p-cymene was used in the synthesis of 5 -isopropyl-8-methylquinoline. It was used as mediator solvent in perchlorate ion-selective electrode with poly(vinyl chloride) matrix membrane on conductive silver-epoxy composite.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Response characteristics of conductive polymer composite substrate all-solid-state poly (vinyl chloride) matrix membrane ion-selective electrodes in aerated and nitrogen-saturated solutions.
Alegret S and Florido A.
Analyst, 116(5), 473-476 (1991)
Reduction of 2-nitro-p-cymene.
Holmes J, et al.
Journal of Chemical and Engineering Data, 27(4), 470-472 (1982)
Preparation of Aldehydes from Alkenes by the Addition of Carbon Monoxide and Hydrogen with Cobalt Carbonyls as Intermediates.
Adkins H and Krsek G.
Journal of the American Chemical Society, 70(1), 383-386 (1948)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service