Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401923

Sigma-Aldrich

Anti-NKD1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Naked1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKD1(85407)

General description

In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin (see MIM 116806)-Tcf (see MIM 602272) signaling pathway.[supplied by OMIM

Immunogen

NKD1 (NP_149110.1, 1 a.a. ~ 470 a.a) full-length human protein.

Sequence
MGKLHSKPAAVCKRRESPEGDSFAVSAAWARKGIEEWIGRQRCPGGVSGPRQLRLAGTIGRSTRELVGDVLRDTLSEEEEDDFRLEVALPPEKTDGLGSGDEKKMERVSEPCPGSKKQLKFEELQCDVSMEEDSRQEWTFTLYDFDNNGKVTREDITSLLHTIYEVVDSSVNHSPTSSKMLRVKLTVAPDGSQSKRSVLVNQADLQSARPRAETKPTEDLRSWEKKQRAPLRFQGDSRLEQSGCYHHCVDENIERRNHYLDLAGIENYTSQFGPGSPSVAQKSELPPRTSNPTRSRSHEPEAIHIPHRKPQGVDPASFHFLDTPIAKVSELQQRLRGTQDGSKHFVRSPKAQGKSVGVGHVARGARNKPPLGPAIPAVSPSAHLAASPALLPSLAPLGHKKHKHRAKESQQGCRGLQAPLASGGPVLGREHLRELPALVVYESQAGQPVQRHEHHHHHEHHHHYHHFYQT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibin Song et al.
EBioMedicine, 35, 155-166 (2018-08-29)
Androgen receptor (AR) plays a crucial role as a transcription factor in promoting the development of hepatocellular carcinoma (HCC) which is prone to aberrant chromatin modifications. However, the regulatory effects of AR on epigenetic mediators in HCC remain ill-defined. Enhancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service