Skip to Content
Merck
All Photos(3)

Documents

SAB1400593

Sigma-Aldrich

Anti-SARS2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FLJ20450, Anti-SARS, Anti-SARSM, Anti-SERS, Anti-SYS, Anti-SerRSmt, Anti-mtSerRS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SARS2(54938)

General description

This gene encodes the mitochondrial seryl-tRNA synthethase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

SARS2 (NP_060297.1, 1 a.a. ~ 518 a.a) full-length human protein.

Sequence
MAASMARRLWPLLTRRGFRPRGGCISNDSPRRSFTTEKRNRNLLYEYAREGYSALPQLDIERFCACPEEAAHALELRKGELRSADLPAIISTWQELRQLQEQIRSLEEEKAAVTEAVRALLANQDSGEVQQDPKYQGLRARGREIRKELVHLYPREAQLEEQFYLQALKLPNQTHPDVPVGDESQARVLHMVGDKPVFSFQPRGHLEIGEKLDIIRQKRLSHVSGHRSYYLRGAGALLQHGLVNFTFNKLLRRGFTPMTVPDLLRGAVFEGCGMTPNANPSQIYNIDPARFKDLNLAGTAEVGLAGYFMDHTVAFRDLPVRMVCSSTCYRAETNTGQEPRGLYRVHHFTKVEMFGVTGPGLEQSSQLLEEFLSLQMEILTELGLHFRVLDMPTQELGLPAYRKFDIEAWMPGRGRFGEVTSASNCTDFQSRRLHIMFQTEAGELQFAHTVNATACAVPRLLIALLESNQQKDGSVLVPPALQSYLGTDRITAPTHVPLQYIGPNQPRKPGLPGQPAVS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service