EP5910752008
Adapter for Eppendorf® S-4x Universal Large Rotor
for 5 mL and 15 mL conical tubes/microtiter and deepwell plates, pkg of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
Recommended Products
feature
Holds 15 mL tubes
for 5 mL and 15 mL conical tubes/microtiter and deepwell plates
packaging
pkg of 2 ea
manufacturer/tradename
Eppendorf® 5910752008
technique(s)
centrifugation: suitable
compatibility
for use with Universal bucket
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service