ABT242
Anti-Galectin-4 Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Galectin-4, Gal-4, Antigen NY-CO-27, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
primate (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), feline (based on 100% sequence homology), monkey (based on 100% sequence homology)
technique(s)
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... LGALS4(3960)
General description
Immunogen
Application
Cell Structure
Adhesion (CAMs)
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Galectin-4 in 10 Āµg of human small intestine tissue lysate.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Galectin-4 in human stomach tissue.
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Galectin-4 in 10 Āµg of HT-29 cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product? Ā
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productās label following the words āLotā or āBatchā.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service