Merck
  • Low expression of SOCS-1 and SOCS-3 is a poor prognostic indicator for gastric cancer patients.
Low expression of SOCS-1 and SOCS-3 is a poor prognostic indicator for gastric cancer patients.
Journal of cancer research and clinical oncology (2014-09-30)
Guanghua Li, Jianbo Xu, Zhao Wang, Yujie Yuan, Yin Li, Shirong Cai, Yulong He
ABSTRACT
Inflammation plays an important role in gastric cancer (GC) development and progression. Suppressor of cytokine signaling (SOCS)-1 and SOCS-3 negatively regulate proinflammatory cytokine signaling; however, their prognostic significance in GC remains unknown. We evaluated the clinicopathological correlation and prognostic significance of SOCS-1 and SOCS-3 in GC. SOCS-1 and SOCS-3 mRNA levels were analyzed in 80 paired gastric tumor and adjacent normal mucosal tissues using a microarray dataset from the Gene Expression Omnibus. Univariate and multivariate analyses were performed to investigate the prognostic impact of SOCS-1 and SOCS-3 immunohistochemical expression on overall survival (OS) and relapse-free survival (RFS) in 186 consecutive GC patients who underwent curative surgery. SOCS-1 and SOCS-3 mRNA expression levels were lower in gastric tumor tissues than in matched normal mucosa. OS and RFS were significantly longer in the high SOCS-1 and SOCS-3 expression groups than in their corresponding low expression groups (p < 0.05). High simultaneous SOCS-1 and SOCS-3 expression were associated with longer OS compared with low simultaneous SOCS-1 and SOCS-3 expression (68.8 vs. 22.2 months; p < 0.001). SOCS-1 [hazards ratio (HR) 0.54, 95 % confidence interval (CI) 0.33-0.87, p = 0.011] and SOCS-3 (HR 0.46, 95 % CI 0.26-0.80, p = 0.006) were independent prognostic factors for OS. Only SOCS-1 (HR 0.20, 95 % CI 0.11-0.38, p = 0.006) was an independent prognostic factor for RFS. Low SOCS-1 and SOCS-3 expression are poor prognostic indicators in GC. GC patients with low SOCS-1 and SOCS-3 expression need close follow-up.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%