Skip to Content
Merck
All Photos(2)

Key Documents

V900516

Sigma-Aldrich

Bicinchoninic acid disodium salt hydrate

≥98% purity , reagent grade, powder

Synonym(s):

2,2′-Biquinoline-4,4′-dicarboxylic acid disodium salt, 4,4′-Dicarboxy-2,2′-biquinoline disodium salt, BCA, Disodium 2,2′-biquinoline-4,4′-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10N2Na2O4 · xH2O
CAS Number:
Molecular Weight:
388.28 (anhydrous basis)
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Product Name

Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

solubility

0.5% sodium bicarbonate: 50 mg/mL, colorless to light yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].[O-]C(=O)c1cc(nc2ccccc12)-c3cc(C([O-])=O)c4ccccc4n3

InChI

1S/C20H12N2O4.2Na.H2O/c23-19(24)13-9-17(21-15-7-3-1-5-11(13)15)18-10-14(20(25)26)12-6-2-4-8-16(12)22-18;;;/h1-10H,(H,23,24)(H,25,26);;;1H2/q;2*+1;/p-2

InChI key

FAFVGCAYKZKOPT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martha Lappas
PloS one, 9(12), e115854-e115854 (2014-12-30)
Infection and inflammation, through their ability to increase pro-inflammatory cytokines and chemokines and adhesion molecules, are thought to play a central role in the pathophysiology of insulin resistance and type 2 diabetes. Recent studies have shown that glycogen synthase kinase
Makon-Sébastien Njock et al.
Blood, 125(20), 3202-3212 (2015-04-04)
The blood contains high concentrations of circulating extracellular vesicles (EVs), and their levels and contents are altered in several disease states, including cardiovascular disease. However, the function of circulating EVs, especially the microRNAs (miRNAs) that they contain, are poorly understood.
Ramya Kumareswaran et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 116(3), 388-391 (2015-04-07)
Solid tumors contain regions of chronic and cyclic hypoxia. Chronic hypoxia can downregulate RAD51 and sensitize cells to PARP inhibition. Herein, we show that RAD51 expression, cell survival and toxicity to PARP inhibition is not affected under cyclic hypoxic conditions.
Naoomi Tominaga et al.
Nature communications, 6, 6716-6716 (2015-04-02)
Brain metastasis is an important cause of mortality in breast cancer patients. A key event during brain metastasis is the migration of cancer cells through blood-brain barrier (BBB). However, the molecular mechanism behind the passage through this natural barrier remains
Fei Zhang et al.
Molecular and cellular endocrinology, 411, 75-85 (2015-04-29)
Anxa2 is dysregulated in many types of carcinomas and implicated in several pivotal biological functions, such as angiogenesis, cell proliferation, invasion, and metastasis. We previously demonstrated that upregulation of Anxa2 enhances the proliferation and invasion of breast cancer cells. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service