Skip to Content
Merck
All Photos(1)

Key Documents

V900926

Sigma-Aldrich

Fulvestrant

Vetec, reagent grade, >98%

Synonym(s):

Fulvestrant, (7α,17β)-7-[9-[(4,4,5,5,5-Pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17-diol, Faslodex, ICI 182,780, ZD 182780, ZD 9238, ZM 182780

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H47F5O3S
CAS Number:
Molecular Weight:
606.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

>98%

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)Cc4cc(O)ccc24

InChI

1S/C32H47F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,21-22,26-29,38-39H,2-10,13-20H2,1H3/t22-,26-,27+,28+,29-,30+,41?/m1/s1

InChI key

VWUXBMIQPBEWFH-WCCTWKNTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Fulvestrant (ICI 182,780) is a selective estrogen receptor down-regulator (SERD). Fulvestrant is a high affinity estrogen receptor antagonist. IC50 = 0.29 nM. Fulvestrant is the first "pure" antiestrogen with no agonistic activity both in vitro and in vivo.
Fulvestrant is a selective estrogen receptor down-regulator (SERD); high affinity estrogen receptor antagonist.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Meng-Yuan Li et al.
Cells, 13(3) (2024-02-09)
(1) Background: Inflammatory responses are implicated in embryo implantation, decidualization, pregnancy maintenance and labor. Both embryo implantation and decidualization are essential to successful pregnancy in rodents and primates. S100A6 is involved in inflammation, tumor development, apoptosis and calcium homeostasis. S100A6
Haimei Li et al.
Life sciences, 344, 122581-122581 (2024-03-22)
Hypocretin is synthesized exclusively in the hypothalamus and distributes inputs to several areas of the brain, which may play an important role in depression. Our previous study showed that hypocretin-1 was increased in the lateral hypothalamus in female patients with
Jieyu He et al.
Aging medicine (Milton (N.S.W)), 6(4), 379-385 (2024-01-19)
To investigate the relationship between icariin and the osteoblastic differentiation of vascular smooth muscle cells (VSMCs) and the signal pathway involved. We applied a universally accepted calcification model of VSMCs induced by β glycerophosphate. Then the VSMCs calcification was observed
Yuet-Kin Leung et al.
PloS one, 9(5), e98037-e98037 (2014-05-20)
Fulvestrant (ICI-182,780) has recently been shown to effectively suppress prostate cancer cell growth in vitro and in vivo. But it is unclear whether microRNAs play a role in regulating oncogene expression in fulvestrant-treated prostate cancer. Here, this study reports hsa-miR-765
Céline Gérard et al.
Oncotarget, 6(19), 17621-17636 (2015-06-10)
Increased risk of breast cancer is a critical side effect associated with the use of a menopausal hormone therapy (MHT). Estetrol (E4) is a natural estrogen produced by the human fetal liver and is a promising compound for clinical use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service