Skip to Content
Merck
All Photos(1)

Key Documents

V900694

Sigma-Aldrich

Trimesic acid

Vetec, reagent grade, 94%

Synonym(s):

Benzene-1,3,5-tricarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CO2H)3
CAS Number:
Molecular Weight:
210.14
Beilstein:
2053080
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

94%

mp

>300 °C (lit.)

SMILES string

OC(=O)c1cc(cc(c1)C(O)=O)C(O)=O

InChI

1S/C9H6O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H,(H,10,11)(H,12,13)(H,14,15)

InChI key

QMKYBPDZANOJGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Long Jiang et al.
Inorganic chemistry, 49(21), 10001-10006 (2010-10-12)
A series of microporous lanthanide-organic framework enantiomers, Ln(BTC)(H(2)O)·(DMF)(1.1) (Ln = Y 1a, 1b; Tb 2a, 2b; Dy 3a, 3b; Er 4a, 4b; Yb 5a, 5b, BTC = 1,3,5-benzenetricarboxylate; DMF = N,N-dimethylformamide) with unprecedented (6,6)-connected topology have been prepared and characterized.
Lin-Hua Xie et al.
Inorganic chemistry, 49(3), 1158-1165 (2010-01-07)
Solvothermal reactions of equimolar zinc acetate, lithium acetate, and 1,3,5-benzenetricarboxylic acid (H(3)btc) in different mixed solvents yielded isostructural three-dimensional frameworks [LiZn(btc)(cG)].lG [cG and lG denote coordinated and lattice guests, respectively; cG = (nmp)(0.5)(H(2)O)(0.5), lG = (EtOH)(0.5) (1a); cG = H(2)O
Leslie J Murray et al.
Journal of the American Chemical Society, 132(23), 7856-7857 (2010-05-21)
Reaction of Cr(CO)(6) with trimesic acid in DMF affords the metal-organic framework Cr(3)(BTC)(2).nDMF (BTC(3-) = 1,3,5-benzenetricarboxylate), which is isostructural to Cu(3)(BTC)(2).3H(2)O. Exchanging DMF for methanol and heating at 160 degrees C under dynamic vacuum for 48 h results in the
Charles E Mays et al.
Biomaterials, 33(28), 6808-6822 (2012-07-04)
Quinacrine and related heterocyclic compounds have antiprion activity. Since the infectious pathogen of prion diseases is composed of multimeric PrP(Sc) assemblies, we hypothesized that this antiprion property could be enhanced by attaching multiple quinacrine-derived chloroquinoline or acridine moieties to a
Tarek Baati et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(23), 2311-2314 (2011-07-06)
The quantification of trimesic acid, a constitutive organic linker from the biodegradable porous iron(III) trimesate MIL-100(Fe) (MIL stands for Materials from Institut Lavoisier), has been performed in different biological complex media (liver, spleen and urine) using a liquid-liquid extraction procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service