Skip to Content
Merck
All Photos(3)

Key Documents

V900452

Sigma-Aldrich

Hypoxanthine

Vetec, reagent grade, 98%

Synonym(s):

6-Hydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Beilstein:
5811
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

mp

>300 °C (lit.)

solubility

formic acid: water (2:1): 50 mg/mL, clear to slightly hazy

SMILES string

O=C1NC=Nc2nc[nH]c12

InChI

1S/C5H4N4O/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)

InChI key

FDGQSTZJBFJUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hypoxanthine is a nutrient additive for a variety of cell culture applications involving bacterial, parasite (Plasmodium falciparum) and animal cells. Hypoxanthine is a component of selection media used in hybridoma technologies.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyu Yu et al.
iScience, 26(1), 105725-105725 (2022-12-30)
The emergence and spread of artemisinin-tolerant malaria parasites threatens malaria control programmes worldwide. Mutations in the propeller domain of the Kelch13 protein confer Plasmodium falciparum artemisinin resistance (ART-R). ART-R is linked to the reduced susceptibility of temporary growth-arrested ring-stage parasites
Chien-Wei Hou et al.
The American journal of Chinese medicine, 40(5), 979-991 (2012-08-30)
Hyperuricemia causes gouty arthritis, kidney disease, heart disease, and other diseases. Xanthine oxidase (XOD) and urate transporters play important roles in urate homeostasis. Numerous plants have been identified as XOD inhibitors. Longan seeds are known to contain high levels of
Tomáš Tichý et al.
European journal of medicinal chemistry, 55, 307-314 (2012-08-04)
Hexadecyloxypropyl esters of acyclic nucleoside phosphonates containing guanine (G) or hypoxanthine (Hx) and a (S)-[3-hydroxy-2-(phosphonomethoxy)propyl] [(S)-HPMP] or 2-(2-phosphonoethoxy)ethyl (PEE) acyclic moiety have been prepared. The activity of the prodrugs was evaluated in vitro against different virus families. Whereas ester derivatives of
Abdulazeez T Lawal et al.
Food chemistry, 135(4), 2982-2987 (2012-09-18)
A potentiometric enzyme electrode for detection of hypoxanthine (Hx) in fish meat is described. The sensor was developed by entrapment of xanthine oxidase (XOD) and ferrocene carboxylic acid (Fc) into polypyrrole (PPy) film during galvanostatic polymerisation film formation. The responses
Chia-Tung Wu et al.
Cardiovascular research, 102(3), 487-496 (2014-03-07)
Fibroblasts, which play an important role in cardiac function/dysfunction, including arrhythmogenesis, have voltage-dependent (Kv) currents of unknown importance. Here, we assessed the differential expression of Kv currents between atrial and ventricular fibroblasts from control dogs and dogs with an atrial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service