Skip to Content
Merck
All Photos(3)

Key Documents

V900435

Sigma-Aldrich

D-(+)-Maltose monohydrate

Vetec, reagent grade

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

reagent grade

product line

Vetec

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dengjun He et al.
Journal of separation science, 38(13), 2229-2237 (2015-04-16)
Trehalose, a nonreducing disaccharide, has been extensively applied to food, cosmetics, and pharmaceutical goods. The resultant solution of trehalose prepared by enzymatic methods includes high amounts of maltose. However, it is quite difficult to separate maltose and trehalose on an
Li Guo et al.
Carbohydrate polymers, 128, 154-162 (2015-05-26)
To understand the susceptibility of glutinous rice starch to digestive enzymes and its potential impact on glycemic response, enzyme kinetics and in vitro digestibility of the native and gelatinized starches were investigated. The results showed that the Km values of
Sanja Milkovska-Stamenova et al.
Journal of agricultural and food chemistry, 63(25), 5911-5919 (2015-06-06)
Glycation is a ubiquitous nonenzymatic reaction of carbonyl compounds with amino groups of peptides and proteins, resulting in the formation of advanced glycation end-products (AGEs) and thereby affecting the properties and quality of thermally processed foods. In this context, mechanisms
Elodie Alinat et al.
Electrophoresis, 36(14), 1555-1563 (2015-03-31)
This work focuses on the development of a CE method allowing, for the first time, the simultaneous separation of the underivatized first seven cellodextrin oligomers (glucose, cellobiose, cellotriose, cellotetraose, cellopentaose, cellohexaose, and celloheptaose), with a view to analyze the hydrolysates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service