Skip to Content
Merck
All Photos(3)

Key Documents

V900381

Sigma-Aldrich

MES hydrate

Vetec, reagent grade, ≥99%

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid hydrate, 4-Morpholineethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4S · xH2O
CAS Number:
Molecular Weight:
195.24 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

form

powder

useful pH range

5.5-6.7

pKa 

6.1

solubility

water: 0.25 g/mL, clear, colorless

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyoshi Shirai et al.
Journal of molecular biology, 321(5), 879-889 (2002-09-11)
The crystal structure of congerin II, a galectin family lectin from conger eel, was determined at 1.45A resolution. The previously determined structure of its isoform, congerin I, had revealed a fold evolution via strand swap; however, the structure of congerin
Jin-Peng Sun et al.
The Journal of biological chemistry, 278(14), 12406-12414 (2003-01-28)
Protein-tyrosine phosphatase 1B (PTP1B) has been implicated as an important regulator in several signaling pathways including those initiated by insulin and leptin. Potent and specific PTP1B inhibitors could serve as useful tools in elucidating the physiological functions of PTP1B and
Christian Engelbrekt et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(13), 2844-2853 (2010-08-18)
Monodisperse platinum nanoparticles (PtNPs) were synthesized by a green recipe. Glucose serves as a reducing agent and starch as a stabilization agent to protect the freshly formed PtNP cores in buffered aqueous solutions. Among the ten buffers studied, 2-(N-morpholino)ethanesulfonic acid
Pasi S Vuorinen et al.
Journal of chromatography. A, 990(1-2), 45-52 (2003-04-11)
A contactless conductivity detector integrated into the capillary cassette of Agilent (3D)CE equipment is described. The detector is user-friendly, compact and easily modified. The UV detector of the (3D)CE equipment is available parallel with the contactless conductivity detector increasing the
Elisabet Fuguet et al.
Electrophoresis, 29(13), 2841-2851 (2008-06-12)
The performance of the most common and also some other less common CE buffers has been tested for the pKa determination of several types of compounds (pyridine, amines, and phenols). The selected buffers cover a pH ranging from 3.7 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service