Skip to Content
Merck
All Photos(2)

Key Documents

V900328

Sigma-Aldrich

Sodium phosphate monobasic dihydrate

Vetec, reagent grade, ≥99%

Synonym(s):

Sodium dihydrogen phosphate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

≥99%

SMILES string

O.O.[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+1;;;/p-1

InChI key

VBJGJHBYWREJQD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bin-Hong Tsai et al.
Polymers, 12(6) (2020-06-19)
Hydrogenated styrenic block copolymers (HSBCs) have been used in medical tubing for many years due to their high clarity, flexibility, kink resistance, and toughness. However, when it comes to blood storage applications, HSBC compounds' market has been limited because of
Sercan Yıldırım et al.
Biomedical chromatography : BMC, 34(10), e4925-e4925 (2020-06-20)
Concentrations of fluoroquinolones, which are used in the treatment of many bacterial infections, should be monitored in biological fluids as they exhibit concentration-dependent bactericidal activity. In this study, a liquid chromatography method for the determination of levofloxacin, ciprofloxacin, moxifloxacin and
Deniz Turan et al.
Food & function, 9(4), 2193-2200 (2018-03-16)
The conjugation reaction of electrospun fibers of a mixture of whey protein isolate (WPI) and dextran using different molecular weights (40, 70, and 100 kDa) and mixing ratios was studied. This study includes the electrospinnability of a mixture of WPI
Matthew P Sullivan et al.
Electrophoresis, 40(18-19), 2329-2335 (2019-05-16)
Cisplatin and its second and third generation analogues are widely used in the treatment of cancer. To study their reactions with proteins, we present a method based on SDS-PAGE separation and laser ablation-inductively coupled plasma-mass spectrometry (LA-ICP-MS) for platinum detection
Anja Schlosser et al.
Physical chemistry chemical physics : PCCP, 21(18), 9002-9012 (2019-03-07)
Semiconductor nanoparticle based porous 3D assemblies are interesting materials for various applications in the fields of photovoltaics, catalysis, or optical sensing. For use as photoelectrodes in photoelectrochemical sensors they need to be characterised by a high porosity, a good photostability

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service