V900073
DL-Malic acid
Vetec™, reagent grade, 98%
Synonym(s):
(±)-2-Hydroxysuccinic acid, DL-Hydroxybutanedioic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein:
1723539
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
reagent grade
vapor density
4.6 (vs air)
vapor pressure
<0.1 mmHg ( 20 °C)
product line
Vetec™
Assay
98%
autoignition temp.
644 °F
mp
131-133 °C (lit.)
SMILES string
OC(CC(O)=O)C(O)=O
InChI
1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)
InChI key
BJEPYKJPYRNKOW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
397.4 °F
Flash Point(C)
203 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
G Anneroth et al.
Scandinavian journal of dental research, 88(3), 214-218 (1980-06-01)
Two saliva stimulating drugs, solutions of Hybrin and malic acid were in an in vitro study found to have a heavy demineralizing effect on the root surface of the dental hard tissue. Consequently the drugs cannot be recommended to patients
Christoph Knuf et al.
Applied and environmental microbiology, 79(19), 6050-6058 (2013-07-31)
Malic acid has great potential for replacing petrochemical building blocks in the future. For this application, high yields, rates, and titers are essential in order to sustain a viable biotechnological production process. Natural high-capacity malic acid producers like the malic
Cuiyue Liang et al.
Plant physiology, 161(3), 1347-1361 (2013-01-24)
Low pH, aluminum (Al) toxicity, and low phosphorus (P) often coexist and are heterogeneously distributed in acid soils. To date, the underlying mechanisms of crop adaptation to these multiple factors on acid soils remain poorly understood. In this study, we
Hugo Oliveira et al.
PloS one, 9(10), e108376-e108376 (2014-10-08)
Resistance rates are increasing among several problematic Gram-negative pathogens, a fact that has encouraged the development of new antimicrobial agents. This paper characterizes a Salmonella phage endolysin (Lys68) and demonstrates its potential antimicrobial effectiveness when combined with organic acids towards
Rafael Moreno-Sánchez et al.
The FEBS journal, 280(3), 927-938 (2012-12-05)
Succinate-driven oxidation via complex II (CII) may have a significant contribution towards the high rates of production of reactive oxygen species (ROS) by mitochondria. Here, we show that the CII Q site inhibitor thenoyltrifluoroacetone (TTFA) blocks succinate + rotenone-driven ROS
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service