PHL23737
Thymoquinone

phyproof® Reference Substance
Synonym(s):
Thymoquin, 2-Isopropyl-5-methyl-1,4-benzoquinone
About This Item
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥95.0% (HPLC)
technique(s)
HPLC: suitable
bp
230-232 °C (lit.)
mp
45-47 °C (lit.)
application(s)
food and beverages
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
CC(C)C1=CC(=O)C(C)=CC1=O
InChI
1S/C10H12O2/c1-6(2)8-5-9(11)7(3)4-10(8)12/h4-6H,1-3H3
InChI key
KEQHJBNSCLWCAE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service