Skip to Content
Merck
All Photos(1)

Key Documents

U1881

Sigma-Aldrich

Uridine 5′-diphosphoglucose-[glucose-6-3H] ammonium salt

aqueous ethanol solution

Synonym(s):

UDP-Glucose, UDP-glc, UDPG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H22N2O17P2 · NH3
CAS Number:
Molecular Weight:
581.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous ethanol solution

extent of labeling

3-10 Ci/mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

N.[H]C(O)C1OC(OP(O)(=O)OP(O)(=O)OCC2OC(C(O)C2O)N3C=CC(=O)NC3=O)C(O)C(O)C1O

Physical form

Ethanol: water (1:1) solution

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey A Gross et al.
BMC genomics, 16, 672-672 (2015-09-04)
The recent discovery that methylated cytosines are converted to 5-hydroxymethylated cytosines (5hmC) by the family of ten-eleven translocation enzymes has sparked significant interest on the genomic location, the abundance in different tissues, the putative functions, and the stability of this
Hiroshi Fukayama et al.
Bioscience, biotechnology, and biochemistry, 78(4), 609-613 (2014-07-19)
Phosphoenolpyruvate carboxylase (PEPC) undergoes activity regulation through reversible phosphorylation. The day/night phosphorylation of leaf PEPC in 27 C3 plant species was analyzed by immunoblotting. PEPC was phosphorylated in the daytime in 12 species, whereas it was phosphorylated at night in
Susannah M L Gagnon et al.
The Journal of biological chemistry, 290(45), 27040-27052 (2015-09-17)
Homologous glycosyltransferases α-(1→3)-N-acetylgalactosaminyltransferase (GTA) and α-(1→3)-galactosyltransferase (GTB) catalyze the final step in ABO(H) blood group A and B antigen synthesis through sugar transfer from activated donor to the H antigen acceptor. These enzymes have a GT-A fold type with characteristic
Robyn Meech et al.
Molecular pharmacology, 87(3), 442-450 (2014-12-19)
The human UDP glycosyltransferase (UGT) superfamily comprises four families of enzymes that catalyze the addition of sugar residues to small lipophilic chemicals. The UGT1 and UGT2 enzymes use UDP-glucuronic acid, and UGT3 enzymes use UDP-N-acetylglucosamine, UDP-glucose, and UDP-xylose to conjugate
Karen M J Saerens et al.
FEMS yeast research, 15(7) (2015-08-25)
Altering glycolipid structure by genetic engineering of Starmerella bombicola is a recently started research topic and worthy alternative to the unsuccessful selective feeding strategies conventionally applied to reach this goal. One question to be addressed when expressing heterologous proteins in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service