Skip to Content
Merck
All Photos(2)

Key Documents

SAB1400487

Sigma-Aldrich

Anti-SPAG8 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-BS-84, Anti-HSD-1, Anti-MGC26201, Anti-SMP1, Anti-SPAG3, Anti-hSMP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
human
citations:
technique(s):
western blot: 1 μg/mL

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPAG8(26206)

General description

The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein encoded by this gene is recognized by sperm agglutinating antibodies from an infertile woman. This protein is localized in germ cells of the testis at all stages of spermatogenesis and is localized to the acrosomal region of mature spermatozoa. Alternatively spliced variants that encode different protein isoforms have been described but the full-length sequences of only two have been determined. (provided by RefSeq)

Immunogen

SPAG8 (AAH19247.1, 1 a.a. ~ 501 a.a) full-length human protein.

Sequence
METNGSTEGSRSRSRSLDIQPSSEGLGPTSEPFPSSDDSPRSALAAATAAAAAAASAAAATAAFTTAKAAALSTKTPAPCSEFMEPSSDPSLLGEPCAGPGFTHNIAHGSLGFEPVYVSCIAQDTCTTTDHSSNPGPVPGSSSGPVLGSSSGAGHGSGSGSGPGCGSVPGSGSGPGPGSGPGSGPGHGSGSHPGPASGPGPDTGPDSELSPCIPPGFRNLVADRVLNYTSWSQHCPWEPQKQPPWEFLQVLEPGARGLWKPPDIKGKLMVCYETLPRGQCLLYNWEEERATNHLDQVPSMQDGSESFFFRHGHRGLLTMQLKSPMPSSTTQKDSYQPPGNVYWPLRGKREAMLEMLLQHQICKEVQAEQEPTRKLFEVESVTHHDYRMELAQAGTPAPTKPHDYRQEQPETFWIQRAPQLPTWWPLPTQVPAAEDYLTWKEWGFTGVQEVLSALLRATPGEYSVNICGMNEHPVCSRTWTNRLCHQEMGSKKTVTQEDRGW

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service