Skip to Content
Merck
All Photos(3)

Key Documents

SAB1400313

Sigma-Aldrich

Anti-PARG antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PARG99

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARG(8505)

General description

The gene encoding PARG is localized on human chromosome 10q11.23. The enzyme has both endo- and exoglycosidase activities and exists as four isoforms.

Immunogen

PARG (NP_003622.2, 1 a.a. ~ 976 a.a) full-length human protein.

Sequence
MNAGPGCEPCTKRPRWGAATTSPAASDARSFPSRQRRVLDPKDAHVQFRVPPSSPACVPGRAGQHRGSATSLVFKQKTITSWMDTKGIKTAESESLDSKENNNTRIESMMSSVQKDNFYQHNVEKLENVSQLSLDKSPTEKSTQYLNQHQTAAMCKWQNEGKHTEQLLESEPQTVTLVPEQFSNANIDRSPQNDDHSDTDSEENRDNQQFLTTVKLANAKQTTEDEQAREAKSHQKCSKSCDPGEDCASCQQDEIDVVPESPLSDVGSEDVGTGPKNDNKLTRQESCLGNSPPFEKESEPESPMDVDNSKNSCQDSEADEETSPGFDEQEDGSSSQTANKPSRFQARDADIEFRKRYSTKGGEVRLHFQFEGGESRTGMNDLNAKLPGNISSLNVECRNSKQHGKKDSKITDHFMRLPKAEDRRKEQWETKHQRTERKIPKYVPPHLSPDKKWLGTPIEEMRRMPRCGIRLPLLRPSANHTVTIRVDLLRAGEVPKPFPTHYKDLWDNKHVKMPCSEQNLYPVEDENGERTAGSRWELIQTALLNKFTRPQNLKDAILKYNVAYSKKWDFTALIDFWDKVLEEAEAQHLYQSILPDMVKIALCLPNICTQPIPLLKQKMNHSITMSQEQIASLLANAFFCTFPRRNAKMKSEYSSYPDINFNRLFEGRSSRKPEKLKTLFCYFRRVTEKKPTGLVTFTRQSLEDFPEWERCEKPLTRLHVTYEGTIEENGQGMLQVDFANRFVGGGVTSAGLVQEEIRFLINPELIISRLFTEVLDHNECLIITGTEQYSEYTGYAETYRWSRSHEDGSERDDWQRRCTEIVAIDALHFRRYLDQFVPEKMRRELNKAYCGFLRPGVSSENLSAVATGNWGCGAFGGDARLKALIQILAAAAAERDVVYFTFGDSELMRDIYSMHIFLTERKLTVGDVYKLLLRYYNEECRNCSTPGPDIKLYPFIYHAVESCAETADHSGQRTGT

Biochem/physiol Actions

PARG (Poly ADP-ribose glycohydrolase) is involved in several gene regulation processes such as maintenance of chromatin structure, transcription, DNA repair and apoptosis. It regulates retinoic acid receptor (RAR)-mediated transcription during chromatin remodeling. It catabolically separate ADP-ribose polymers synthesized by poly-(ADP-ribose) polymerases. A study shows that PARG plays a therapeutic role in various physiological conditions such as inflammation, ischemia, stroke, and cancer chemotherapy.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly(ADP-ribosyl) glycohydrolase prevents the accumulation of unusual replication structures during unperturbed S phase.
Ray Chaudhuri A
Molecular and Cellular Biology (2015)
Human poly (ADP-ribose) glycohydrolase (PARG) gene and the common promoter sequence it shares with inner mitochondrial membrane translocase 23 (TIM23)
Ralph G Meyer
Gene, 181-190 (2003)
Wookee Min et al.
Frontiers in bioscience (Landmark edition), 14, 1619-1626 (2009-03-11)
Poly (ADP-robose) glycohydrolase (PARG) is a catabolic enzyme that cleaves ADP-ribose polymers synthesized by members of the poly (ADP-ribose) polymerase (PARP) family of enzymes. The growing evidence supports the importance of a tight control of poly (ADP-ribose) metabolism by the
M-E Bonicalzi et al.
Cellular and molecular life sciences : CMLS, 62(7-8), 739-750 (2005-05-04)
Poly(ADP-ribose) glycohydrolase (PARG) is a catabolic enzyme that cleaves ADP-ribose polymers formed by members of the PARP family of enzymes. Despite its discovery and subsequent partial purification in the 1970s and the cloning of its single gene in the late
Nicolas Le May et al.
Molecular cell, 48(5), 785-798 (2012-10-30)
Poly-(ADP-ribose) glycohydrolase (PARG) is a catabolic enzyme that cleaves ADP-ribose polymers synthesized by poly-(ADP-ribose) polymerases. Here, transcriptome profiling and differentiation assay revealed a requirement of PARG for retinoic acid receptor (RAR)-mediated transcription. Mechanistically, PARG accumulates early at promoters of RAR-responsive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service