Skip to Content
Merck
All Photos(3)

Documents

HPA001599

Sigma-Aldrich

Anti-MXD1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MAD protein antibody produced in rabbit, Anti-MAX dimerization protein 1 antibody produced in rabbit, Anti-MAX dimerizer antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LCLEKLKGLVPLGPESSRHTTLSLLTKAKLHIKKLEDCDRKAVHQIDQLQREQRHLKRQLEKLGIERIRMDSIGSTVSSERSDSDREEIDVDVESTDYLTGDLDWSSSSVSDSDERGSMQSLGSDEGYSSTSI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MXD1(4084)

General description

MXD1 (MAX dimerization protein 1) is a basic helix-loop helix-leucine-zipper (bHLHZip) transcription suppressor, which belongs to the MYC/MAX/MAD family of transcription factors. Its N-terminal contains a Sin3-interacting domain (SID). MAD family consists of four members namely, MAD1 (MXD1), MXI1, MAD3 and MAD4.

Immunogen

MAD protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MXD1 (MAX dimerization protein 1) functions as an antagonist of Myc/Max complex by competing for common DNA targets and recruiting repressor complexes containing histone deacetylases. Defects in this gene is associated with acute leukemia. It also acts as a tumor suppressor. It suppresses the transcription of human telomerase (hTERT) gene in mortal cells. The antagonism of MXD1 by Myc might be one of the mechanisms resulting in immortal cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84736

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian A Schardt et al.
Methods in enzymology, 489, 227-243 (2011-01-27)
There is accumulating evidence for the involvement of the unfolded protein response (UPR) in the pathogenesis of many tumor types in humans. This is particularly the case in rapidly growing solid tumors in which the demand for oxygen and nutrients
Adam G Sowalsky et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(14), 3823-3833 (2017-01-26)
Purpose: The molecular features that account for the distinct histology and aggressive biological behavior of Gleason pattern 4 (Gp4) versus Gp3 prostate cancer, and whether Gp3 tumors progress directly to Gp4, remain to be established.Experimental Design: Whole-exome sequencing and transcriptome
B Amati et al.
Current opinion in genetics & development, 4(1), 102-108 (1994-02-01)
The Myc oncoprotein dimerizes with its partner, Max, to bind DNA, activate transcription, and promote cell proliferation, as well as programmed cell death. Max also forms homodimers or heterodimers with its alternative partners, Mad and Mxi-1. These complexes behave as
A L Eilers et al.
The Journal of biological chemistry, 274(46), 32750-32756 (1999-11-07)
Members of the Mad family of bHLHZip proteins heterodimerize with Max and function to repress the transcriptional and transforming activities of the Myc proto-oncogene. Mad:Max heterodimers repress transcription by recruiting a large multi-protein complex containing the histone deacetylases, HDAC1 and
S Oh et al.
Oncogene, 19(11), 1485-1490 (2000-03-21)
Activation of telomerase, which has been frequently associated with cellular immortality, may constitute a key step in the development of human cancer. De-repression in the expression of its catalytic subunit hTERT gene has been proposed to directly link to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service