AXYQT1100CBKRS
Pipette tips for Qiagen®
capacity 1100 μL, black, racked, pkg of 16x96tips/cs, sterile
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1100 μL
sterility:
sterile
packaging:
pkg of 16x96tips/cs
manufacturer/tradename:
Axygen Inc QT-1100-CBK-R-S
Corning
Corning
sterility
sterile
manufacturer/tradename
Axygen Inc QT-1100-CBK-R-S
Corning
packaging
pkg of 16x96tips/cs
capacity
1100 μL
volume
1100 μL
color
black
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Legal Information
Qiagen is a registered trademark of Qiagen Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service