Skip to Content
Merck
All Photos(1)

Documents

AV33478

Sigma-Aldrich

Anti-UBP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Upstream binding protein 1 (LBP-1a)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

60 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBP1(7342)

General description

UBP1 is a transcription factor shown to effect expression of HIV Type 1.

Immunogen

Synthetic peptide directed towards the middle region of human UBP1

Biochem/physiol Actions

UBP1 is an important SF1-independent transcriptional activator stimulating P450scc expression in human placental JEG-3 cells, whereas LBP-9 modulates the action of UBP1, exerting both positive and negative effects

Sequence

Synthetic peptide located within the following region: GASQTSGEQIQPSATIQETQQWLLKNRFSSYTRLFSNFSGADLLKLTKED

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Yoon et al.
Molecular and cellular biology, 14(3), 1776-1785 (1994-03-01)
LBP-1 is a cellular protein which binds strongly to sequences around the human immunodeficiency virus type 1 (HIV-1) initiation site and weakly over the TATA box. We have previously shown that LBP-1 represses HIV-1 transcription by inhibiting the binding of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service