Pricing and availability is not currently available.
Assay
≥90% (coupling to amines)
manufacturer/tradename
ATTO-TEC GmbH
technique(s)
UV/Vis spectroscopy: suitable
fluorescence
λem 615 nm
suitability
corresponds for UV test suitable for fluorescence
storage temp.
−20°C
Application
Atto 612Q is a quencher for fluorescence characterized by high molecular absorption (105.000). Atto 612Q is characterized by a high photostability and thermostability. It is moderately hydrophilic, and soluble in polar solvents as DMF or DMSO.
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
We describe the synthesis and properties of new fluorescence quenchers containing aldehyde, hydrazine, and aminooxy groups, allowing convenient bioconjugation as oximes or hydrazones. Conjugation to oligonucleotides proceeded in high yield with aniline as catalyst. Kinetics studies of conjugation show that
Physical chemistry chemical physics : PCCP, 13(21), 9929-9941 (2011-03-08)
Fluorescence-based assays and detection techniques are among the most highly sensitive and popular biological tests for researchers. To match the needs of research and the clinic, detection limits and specificities need to improve, however. One mechanism is to decrease non-specific
Journal of the American Chemical Society, 132(43), 15418-15426 (2010-10-14)
In the development of orthogonal extra base pairs for expanding the genetic alphabet, we created novel, unnatural base pairs between fluorophore and quencher nucleobase analogues. We found that the nucleobase analogue, 2-nitropyrrole (denoted by Pn), and its 4-substitutions, such as
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.