Skip to Content
Merck
All Photos(1)

Key Documents

40592

Sigma-Aldrich

Atto 540 Q

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116100
NACRES:
NA.32
Pricing and availability is not currently available.

product line

BioReagent

Assay

≥98.0% (HPCE)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

technique(s)

UV/Vis spectroscopy: suitable

suitability

corresponds for UV test
suitable for fluorescence

storage temp.

−20°C

General description

Atto 540Q is a new quencher for fluorescence characterized by high molecular absorption (105.000). Atto 540Q is characterized by a high photostability and thermostability. It is moderately hydrophilic, and soluble in polar solvents as DMF or DMSO.

Application

Atto 540 Q is suitable for preparation of fluorescence quenching reagents and the study of its physicochemical properties.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service