Skip to Content
Merck
All Photos(1)

Key Documents

341M-9

Sigma-Aldrich

TRAcP (9C5) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

9C5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (341M-94)
vial of 0.5 mL concentrate (341M-95)
bottle of 1.0 mL predilute (341M-97)
vial of 1.0 mL concentrate (341M-96)
bottle of 7.0 mL predilute (341M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2b

control

hairy cell leukemia

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... ACP5(54)

General description

Tartrate resistant acid phosphatase (TRAcP) is a basic, iron-binding protein with high activity towards phosphoproteins, ATP and 4 nitrophenyl phosphate. [1][2][3][4] Expression of TRAcP is reported to be increased in the spleen and monocytes of individuals with Gaucher′s disease, splenocytes and circulating white cells of individuals with hairy cell leukemia, spleens of individuals with Hodgkin disease, and the sera of individuals undergoing active bone turnover. [1][2][3] Elevated levels are also reported to be associated with various B-cell and T-cell leukemias and lymphomas, placental decidual cells, syncytiotrophoblasts, and some macrophages distributed throughout maternal and embryonic tissues. [1][2][3][4]

The histochemical identification of hairy cell leukemia via tartrate-resistant acid phosphatase assay has been a standard for over two decades. Anti-TRAcP labels the cells of hairy cell leukemia (HCL) with a high degree of sensitivity and specificity. Worthy also of mention in this regard are anti-annexin A1 and anti-CD11c. Other cells stained with anti-TRAcP are tissue macrophages and osteoclasts, which also express abundant TRAcP activity. [1][2][3][4][5]

Quality


IVD

IVD

IVD

RUO

Linkage

TRAcP Positive Control Slides, Product No. 341S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Yaziji et al.
American journal of clinical pathology, 104(4), 397-402 (1995-10-01)
Immunohistochemical studies were done on formalin-fixed, paraffin-embedded tissues to evaluate the specificity of a newly developed monoclonal antibody (9C5) against tartrate-resistant acid phosphatase. Sections from 195 specimens were examined, which included 33 types of tissues/organs. These tissues included normal, inflammatory
J D Hoyer et al.
American journal of clinical pathology, 108(3), 308-315 (1997-09-18)
The demonstration of tartrate-resistant acid phosphatase (TRAP) activity has long been a cornerstone in the diagnosis of hairy cell leukemia (HCL). Recently a monoclonal antibody to this enzyme has been developed that can be used in an immunoperoxidase method on
A J Janckila et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 44(3), 235-244 (1996-03-01)
We have developed a monoclonal antibody (9C5) for immunohistochemical localization of tartrate-resistant acid phosphatase (TRAcP). This antibody reacts with a denatured epitope of TRAcP and requires enhancement methods to promote antigenicity in paraffin-embedded tissues. We used this antibody to systematically
A J Janckila et al.
Blood, 85(10), 2839-2844 (1995-05-15)
Tartrate-resistant acid phosphatase (TRAcP) has been an indispensible marker for hairy cell leukemia (HCL) for over two decades. However, the traditional TRAcP cytochemical stain cannot be performed effectively on sections of paraffin-embedded tissues that are important resources for histopathologic evaluation
A J Janckila et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 73(6), 316-324 (1999-01-15)
Tartrate-resistant acid phosphatase (TRAP) is expressed abundantly by osteoclasts and is required for bone resorption. This enzyme is emerging as an important biomarker in bone pathology, both for histochemical identification of osteoclasts and as a serum marker of osteoclast activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service