Skip to Content
Merck
All Photos(1)

Documents

14699

Sigma-Aldrich

2-Deoxy-D-ribonic acid lithium salt

≥95.0% (TLC)

Synonym(s):

2-Deoxy-D-erythro-pentonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5 · xLi+
CAS Number:
Molecular Weight:
150.13 (free acid basis)
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (TLC)

optical activity

[α]/D 12.0±2.0°, 24 hr, c = 1 in 1 M HCl

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@@H](O)CC(O)=O

InChI

1S/C5H10O5/c6-2-4(8)3(7)1-5(9)10/h3-4,6-8H,1-2H2,(H,9,10)/t3-,4+/m0/s1

InChI key

VBUWJOHKCBQXNU-IUYQGCFVSA-N

Biochem/physiol Actions

2-Deoxyribonic acid is the acid form of deoxyribonate, intercovertible with 2-Deoxy-ribonolactone, produced as part of bistranded lesions by DNA damaging agents, by ionizing radiation, organometallic oxidants, and is a metastable intermediate in DNA damage mediated by copper phenanthroline nucleases, and is also formed under anaerobic conditions in the presence of the radiosensitizing agent tirapazamine. It has been found in normal human biofluids.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of 2-deoxy-D-erythro-pentonic acid from an alkali-labile site in gamma-irradiated DNA.
M Dizdaroglu et al.
International journal of radiation biology and related studies in physics, chemistry, and medicine, 32(5), 481-483 (1977-11-01)
M Davies et al.
Acta endocrinologica, 104(2), 210-215 (1983-10-01)
Serum vitamin D metabolites, the renal tubular maximum reabsorptive rate for phosphate (TMP/GFR) nephrogenic cyclic AMP (NcAMPI, and CaE (urinary calcium excretion per litre of glomerular filtrate) were measured in 14 adults with familial hypocalciuric hypercalcaemia (FHH). The findings were
Kelly M Kroeger et al.
Biochemistry, 42(8), 2449-2455 (2003-02-26)
2-Deoxyribonolactone (3) is produced in DNA as a result of reaction with a variety of DNA damaging agents. The lesion undergoes beta-elimination to form a second metastable electrophilic product (4). In this study, DNA containing 2-deoxyribonolactone (3) and its beta-elimination
Cleavage of the intermediate hydroperoxides in the oxidation of D-glucose and D-fructose with oxygen.
Vuorinen, T.
Carbohydrate Research, 141, 319-322 (1985)
Formation of carboxylic acids during degradation of mono- saccharides.
Novotny, O., et al.
Czech Journal of Food Sciences, 26, 117-131 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service