Skip to Content
Merck
All Photos(1)

Key Documents

95001

Supelco

N-Debenzoyl-N-hexanoyl-N-methylpaclitaxel

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Paclitaxel impurity F (PhEur), N-Methylpaclitaxel C, N-Methyltaxol C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H59NO14
CAS Number:
Molecular Weight:
861.97
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

CC(O[C@]12[C@@]3([H])[C@H](OC(C4=CC=CC=C4)=O)[C@](C(C)(C)C([C@@H](OC(C)=O)C([C@]3(C)[C@@H](O)C[C@@]1([H])OC2)=O)=C5C)(O)C[C@@H]5OC([C@H](O)[C@@H](N(C(CCCCC)=O)C)C6=CC=CC=C6)=O)=O

InChI

1S/C47H59NO14/c1-9-10-13-22-34(52)48(8)36(29-18-14-11-15-19-29)37(53)43(56)60-31-24-47(57)41(61-42(55)30-20-16-12-17-21-30)39-45(7,32(51)23-33-46(39,25-58-33)62-28(4)50)40(54)38(59-27(3)49)35(26(31)2)44(47,5)6/h11-12,14-21,31-33,36-39,41,51,53,57H,9-10,13

InChI key

LJTMOSWWGSCCPR-AMMYIWEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service