Skip to Content
Merck
All Photos(1)

Key Documents

42505

Millipore

1,4-Dioxane

suitable for HPLC, ≥99.8% (GC)

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

≥99.8% (GC)

autoignition temp.

356 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

22 %

technique(s)

HPLC: suitable

impurities

≤0.05% water

evapn. residue

≤0.001%

refractive index

n20/D 1.422 (lit.)
n20/D 1.422

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

solvent strength ε (Al2O3)

0.56

λ

1 cm path, H2O reference

UV absorption

λ: 220 nm Amax: 1.0
λ: 270 nm Amax: 0.1
λ: 300 nm Amax: 0.01

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service