Skip to Content
Merck
All Photos(1)

Documents

16995

Sigma-Aldrich

(S)-(−)-(2-Bromoethyl)oxirane

technical, ≥90% (sum of enantiomers, GC)

Synonym(s):

(S)-(−)-4-Bromo-1,2-epoxybutane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7BrO
CAS Number:
Molecular Weight:
151.00
Beilstein:
1560144
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (sum of enantiomers, GC)

form

liquid

optical activity

[α]20/D −26±1°, c = 0.3% in chloroform

refractive index

n20/D 1.476

bp

80 °C/50 mmHg (lit.)

density

1.521 g/mL at 20 °C (lit.)

SMILES string

BrCC[C@H]1CO1

InChI

1S/C4H7BrO/c5-2-1-4-3-6-4/h4H,1-3H2/t4-/m0/s1

InChI key

ZKODPGZNBMIZFX-BYPYZUCNSA-N

Other Notes

Versatile bifunctional chiral building block with two different electrophilic centers

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy Hauck Newman et al.
Journal of medicinal chemistry, 52(8), 2559-2570 (2009-04-01)
In the present report, the D3 receptor pharmacophore is modified in the 2,3-diCl- and 2-OCH(3)-phenylpiperazine class of compounds with the goal to improve D3 receptor affinity and selectivity. This extension of structure-activity relationships (SAR) has resulted in the identification of
Y. Yuasa et al.
Synthetic Communications, 27, 573-573 (1997)
D. Seebach et al.
Liebigs Ann. Chem., 1007-1007 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service