Skip to Content
Merck
All Photos(1)

Key Documents

MABX7161

Anti-Cardiac Troponin I (cTnI), in vitro, clone 1611

clone 1611, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1611, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

Gene Information

human ... TNNI3(7137)

General description

Antibody reacts with free cardiac troponin I (cTn1) and cTnI forming complexes with other troponin components. It recognizes epitopes that are not perturbed or sterically shielded by other troponin components

Application

Confirmed matched pairing. Capture Antibody MABX3150-1KC; Detector Antibody MABX7161-1KC

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongpan Huang et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 8836904-8836904 (2020-12-10)
Oxymatrine (OMT) is the primary pharmacological component of Sophora flavescens Aiton., which has been shown to possess potent antifibrotic, antioxidant, and anti-inflammatory activities. The aim of the present study was to clarify the protective mechanism of OMT on acute lung

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service