Skip to Content
Merck
All Photos(2)

Documents

MABS1726

Sigma-Aldrich

Anti-MPK-3 (DUSP6) Antibody, clone 5H6.1

clone 5H6.1, from mouse

Synonym(s):

Dual specificity protein phosphatase 6, Dual specificity protein phosphatase PYST1, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5H6.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DUSP6(1848)

General description

Dual specificity protein phosphatase 6 (EC 3.1.3.16; EC 3.1.3.48; UniProt Q16828; also known as Dual specificity protein phosphatase PYST1, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP-3) is encoded by the DUSP6 (also known as HH19, MKP3, PYST1) gene (Gene ID 1848) in human. MPK-3 (DUSP6) belongs to the family of dual-specificity phosphatases (DUSPs) that mediate dephosphorylation of MAPKs on threonine/serine and tyrosine residues. DUSP6 is upregulated in HER2+ carcinomas and is among several DUSPs thought to be involved in breast cancer metastasis. DUSP1, DUSP4, and DUSP6 are induced during epithelial-to-mesenchymal transition (EMT) and are involved in forming and maintaining breast cancer stem cells (CSCs), DUSP1 knockdown reduces CSC formation, while DUSP4 or DUSP6 knockdown enhances CSC formation. PKC CK2alpha is reported to phosphorylate DUSP6 and increase its dephosphatase activity. ERK1/2, on the other hand, can phosphorylate DUSP6 to promote its proteosomal degradation.

Specificity

Clone 5H6.1 targets an epitope within the rhodanese domain present in both spliced isoforms of human MPK-3 (DUSP6) reported by UniProt (Q16828).

Immunogen

GST-tagged recombinant human MPK-3 (DUSP6) rhodanese domain fragment.

Application

Detect DUSP6 using this mouse monoclonal Anti-MPK-3 (DUSP6), clone 5H6.1 Antibody, Cat. No. MABS1726, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected MPK-3 (DUSP6) in human liver, pancreas, and tonsil tissue sections.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MPK-3 (DUSP6) in 10 µg of HepG2 cell lysate.

Target description

~42 kDa observed. 42.32 kDa (isoform 1) and 26.47 kDa (isoform 2; DUSP6-ALT) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service