Skip to Content
Merck
All Photos(1)

Documents

5096-M

Sigma-Aldrich

Mounting Fluid, Low Glycerol, recommended for Rabies

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405

form

liquid

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

General description

Mounting fluid, low glycerol.

Application

Recommended for use with the following Rabies Reagents: Rabies DFA Reagent (Cat. #5100), Rabies DFA II Reagent (Cat. #5500), and the Rabies Polyclonal Reagent (Cat. #5199).

Physical form

Low glycerol mounting fluid in buffered saline solution with 0.2% sodium azide. Ready to use; do not dilute.

Storage and Stability

When stored at 2-8°C, reagent is stable up to the expiration date printed on the label.

WARNING / PRECAUTION:

Sodium azide, present in this reagent can form potentially explosive metal azides with lead and copper pipes. Upon disposal, flush with large volumes of water to prevent build-up in plumbing.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service