Skip to Content
Merck

5.00878

SAFC

Vitamin E

dry powder, EMPROVE® ESSENTIAL, Ph. Eur., BP

Pharma Manufacturing

Synonym(s):

(±)-α-Tocopherol, Vitamin E dry powder, DL-all-rac-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
94012
MDL number:
UNSPSC Code:
12352401

Agency

BP
Ph. Eur.

Quality Level

product line

EMPROVE® ESSENTIAL

form

dry powder
solid

pH

7 (20 °C, 50 g/L in H2O, slurry)

density

0.950 g/mL at 20 °C (lit.)

application(s)

pharmaceutical

shipped in

ambient

storage temp.

15-25°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3

InChI key

GVJHHUAWPYXKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Vitamin E protects cells against free radicals. It is used in skin and hair care products, as well as in dietary supplements. Vitamin E dry powder sprayed (50 g all-rac-α-tocopheryl acetate in 100 g substance) Emprove® Essential is suitable for pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service