Skip to Content
Merck
All Photos(1)

Key Documents

14-518

Sigma-Aldrich

PKCε Protein, active, 10 µg

Active, N-terminal His6-tagged, recombinant PKCε, amino acids 2-end. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in Sf21 cells

mol wt

Mw 84.6 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... PRKCE(5581)

General description

N-terminal His6-tagged, recombinant PKCε, amino acids 2-end

PKC Lipid activator (Catalog # 20-133) is required for use.
Product Source: Human PKCε expressed in Sf21 insect cells

Biochem/physiol Actions

Protein Target: PKCε

Packaging

2 vials at 5μg each
Also available in 250μg size (2x125μg)--call for pricing and availability and reference catalog number 14-518M when ordering the 250μg size.

Quality

Routinely evaluated by phosphorylation of PKC Substrate Peptide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Zhong Tao et al.
Biochemical and biophysical research communications, 351(3), 708-712 (2006-11-07)
Phosphorylation and O-GlcNAcylation of keratin 18 (K18) are highly dynamic and involve primarily independent K18 populations. We used in vitro phosphorylation and O-GlcNAcylation of wild-type, phospho-Ser52, glyco-Ser48, and Ser-to-Ala mutant 17mer peptides (K18 amino acids 40-56), which include the major
Thomas B Schaffer et al.
Cell reports, 25(9), 2470-2483 (2018-11-30)
Protein kinase C (PKC)-dependent mechanisms promote synaptic function in the mature brain. However, the roles of PKC signaling during synapse development remain largely unknown. Investigating each brain-enriched PKC isoform in early neuronal development, we show that PKCε acutely and specifically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service