Skip to Content
Merck
All Photos(1)

Documents

14-448-M

Sigma-Aldrich

Cdk2/Cyclin A Protein, active, 10 µg

Active, C-terminal His6-tagged human full length Cdk2 & N-terminal GST-tagged human full length Cyclin A, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

product line

Upstate®

specific activity

(For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.)

mol wt

Mw 35 kDa

purified by

(The cdk2 is purified by Ni2+/NTA agarose and then activated using CAK and repurified by Q Sepharose and Ni2+/NTA agarose. The cyclin A is purified using glutathioneagarose)

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

accession no.

 (FUNCTION: SwissProt: P24941 # Involved in the control of the cell cycle. Interacts with cyclins A, B1, B3, D, or E. Activity of CDK2 is maximal during S phase and G2. )
NM_001798.2

NCBI accession no.

UniProt accession no.

General description

C-terminal His6-tagged human full length cdk2, and N-terminal GST-tagged human full length cyclin A

Biochem/physiol Actions

Protein Target: CDK2/cyclinA
Target Sub-Family: CMGC

Packaging

Also available in 10 μg size and in bulk
Also available in 250μg size, please inquire for pricing and availability.

Quality

routinely evaluated by phosphorylation of Histone H1

Target description

CDK2/cyclinA

Linkage

Replaces: 14-407

Physical form

The cdk2 is purified by Ni2+/NTA agarose and then activated using CAK and repurified by Q Sepharose and Ni2+/NTA agarose. The cyclin A is purified using glutathioneagarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

Sepharose is a trademark of Cytiva
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service