Skip to Content
Merck
All Photos(2)

Documents

05-591-AF647

Sigma-Aldrich

Anti-Akt/PKB Antibody, PH Domain, clone SKB1, Alexa Fluor 647

clone SKB1, from mouse, ALEXA FLUOR 647

Synonym(s):

RAC-alpha serine/threonine-protein kinase, PKB, PKB alpha, Protein kinase B, Protein kinase B alpha, Proto-oncogene c-Akt, RAC-PK-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

SKB1, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunocytochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Akt1(11651)

General description

RAC-alpha serine/threonine-protein kinase (EC 2.7.11.1; UniProt P31749; also known as PKB, PKB alpha, Protein kinase B, Protein kinase B alpha, Proto-oncogene c-Akt, RAC-PK-alpha) is encoded by the AKT1 (also known as PKB, RAC) gene (Gene ID 207) in human. AKTs (protein kinase Bs) are serine/threonine kinases that play an important role in signal transduction pathways involved in cell proliferation, apoptosis, angiogenesis, and diabetes. There exit three types of AKTs (α, β, γ or AKT 1, 2, 3) sharing a high degree of homology, but differing slightly in the localization of their regulatory phosphorylation sites. AKT α is the predominant isoform in most tissues, whereas the highest expression of AKT β is observed in the insulin-responsive tissues, and AKT γ is abundant in brain tissue. Each AKT type is composed of three functionally distinct regions, including an N-terminal pleckstrin homology (PH) domain that provides a lipid-binding module to direct Akt to PIP2 and PIP3, a central catalytic domain, and a C-terminal hydrophobic motif.

Specificity

Clone SKB1 recognizes Akt1 and Akt2, but not Akt3, GST fusion proteins at 100 ng by immunoblotting analysis using the unconjugated antibody (Cat. No. 05-591).

Immunogen

Epitope: Amino acids 1-149
GST-fusion protein corresponding to residues 1-149 of human Akt1.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Akt/PKB in A431 cells.
The unconjugated antibody (Cat. No. 05-591) is shown to be suitable also for flow cytometry, immunocytochemistry, immunoprecipitation, and Western blotting applications.
Research Category
Apoptosis & Cancer
Research Sub Category
Kinases & Phosphatases
This Anti-Akt Antibody, PH Domain, clone SKB1, Alexa Fluor 647 is validated for use in Immunocytochemistry for the detection of PKB.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Akt/PKB in HeLa cells.

Target description

~60 kDa observed.

Physical form

Protein G purified.
Purified mouse monoclonal IgG antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service