Skip to Content
Merck
All Photos(1)

Key Documents

790590P

Avanti

Bicelle Blend (DMPC:DHPC (2.8:1, mol:mol))

Avanti Research - A Croda Brand, powder

Synonym(s):

Bicelle Blend (DMPC:DHPC (2.8:1, mol:mol))

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (790590P-200MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Bicelle blend contains 200mg of dimyristoylphosphatidylcholine (DMPC): dihexanoylphosphatidylcholine (DHPC) Pre-Mixed Bicelle (2.8:1, mol:mol). Bicelle is a discoidal lipid aggregate. It is made up of long-chain phospholipid and a detergent. The planar bilayer is generally composed of DMPC, which may have phospholipids with identical chain lengths but differ in the head group. The detergent layer is composed of a short-chain phospholipid like DHPC.
The bicelle blend is designed to aid with the production of high-quality membrane protein crystals. Bicelles can be integrated into standard crystallization protocols, and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation. Reference/Protocol: https://avantilipids.com/tech-support/liposome-preparation/bicelle-preparation/

Biochem/physiol Actions

Bicelles are used as membrane mimetics for studying membrane-associated biomolecules.

Packaging

5ML Amber Glass Screw Cap Vial (790590P-200MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Glover et al.
Biophysical journal, 81(4), 2163-2171 (2001-09-22)
Several complementary physical techniques have been used to characterize the aggregate structures formed in solutions containing dimyristoylphosphatidylcholine (DMPC)/dihexanoylphosphatidylcholine (DHPC) at ratios of < or =0.5 and to establish their morphology and lipid organization as that of bicelles. (31)P NMR studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service