Documents
Z259535
Sigma-Aldrich
Ace photochemical reactor vessel
Complete, capacity 500 mL
About This Item
UNSPSC Code:
41103817
NACRES:
NC.11
manufacturer/tradename
Ace Glass 786338
capacity
500 mL
Related Categories
General description
Borosilicate glass. Complete unit includes vessel, sparger tube, 38 × 8 mm stirrer bar, threaded #7 nylon thermometer bushing, #50 bushing and 3 m of 4.8 mm I.D. FEP tubing.