861901
Lithium dodecyl sulfate
98%
Synonym(s):
Dodecyl lithium sulfate, Dodecyl sulfate lithium salt, Lauryl sulfate lithium salt, Lithium lauryl sulfate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)11OSO3Li
CAS Number:
Molecular Weight:
272.33
Beilstein:
5186718
EC Number:
MDL number:
UNSPSC Code:
12161900
Recommended Products
Assay
98%
CMC
7-10 mM (20-25°C)
SMILES string
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
InChI
1S/C12H26O4S.Li/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1
InChI key
YFVGRULMIQXYNE-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Anionic detergent that may be used in place of SDS for electrophoresis in cold conditions.
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service