All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H8Cl4OP2
CAS Number:
Molecular Weight:
371.95
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
Assay
95%
reaction suitability
reagent type: ligand
mp
62-66 °C
functional group
phosphine
storage temp.
2-8°C
SMILES string
ClP(Cl)c1ccccc1Oc2ccccc2P(Cl)Cl
InChI
1S/C12H8Cl4OP2/c13-18(14)11-7-3-1-5-9(11)17-10-6-2-4-8-12(10)19(15)16/h1-8H
InChI key
XDOGVZTTZQALKT-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service