Skip to Content
Merck
All Photos(1)

Key Documents

669490

Sigma-Aldrich

(1R,4R,8R)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene

95%

Synonym(s):

(1R,4R,8R) Carreira DOLEFIN Ligand, (1R,4R,8R)-8-Methoxy-1,8-dimethyl-2-(2-methylpropyl)-5-(phenylmethyl)bicyclo[2.2.2]octa-2,5-diene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Assay

95%

form

liquid

optical activity

[α]20/D +98±5°, c = 0.7 in chloroform

reaction suitability

reaction type: click chemistry

refractive index

n20/D 1.5205

functional group

ether
phenyl

SMILES string

CO[C@]1(C)C[C@]2(C)C=C(Cc3ccccc3)[C@H]1C=C2CC(C)C

InChI

1S/C22H30O/c1-16(2)11-19-13-20-18(12-17-9-7-6-8-10-17)14-21(19,3)15-22(20,4)23-5/h6-10,13-14,16,20H,11-12,15H2,1-5H3/t20-,21+,22-/m1/s1

InChI key

ACWLDJOHMGJACE-BHIFYINESA-N

Application

(1R, 4R, 8R) Carreira DOLEFIN Ligand is a chiral diene ligand that can be used in:
  • The asymmetric Rh-catalyzed arylation of alkenylheterocycles by conjugate additions with various aryl boronic acids.
  • The enantioselective synthesis of 3,3-diarylpropanals by conjugate addition of aromatic boronic acids to cinnamaldehyde derivatives in the presence of a rhodium catalyst.
  • The asymmetric synthesis of pyrrolizidinones by rhodium-catalyzed 1,4-addition of aryl boronic acids to pyrrolidine acceptor.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium-catalyzed 1, 4-additions to enantiopure acceptors: Asymmetric synthesis of functionalized pyrrolizidinones
Zoute L, et al.
Organic Letters, 11(12), 2491-2494 (2009)
Asymmetric Synthesis of 3, 3-Diarylpropanals with Chiral Diene- Rhodium Catalysts
Paquin J-F, et al.
Journal of the American Chemical Society, 127(31), 10850-10851 (2005)
Enantioselective rhodium-catalyzed addition of arylboronic acids to alkenylheteroarenes
Pattison G, et al.
Journal of the American Chemical Society, 132(41), 14373-14375 (2010)
Jean-François Paquin et al.
Journal of the American Chemical Society, 127(31), 10850-10851 (2005-08-04)
A general route to enantioenriched 3,3-diarylpropanals is presented. These useful building blocks are prepared via an asymmetric rhodium-catalyzed conjugate addition of arylboronic acids to cinnamaldehyde derivatives in the presence of chiral dienes. The addition of both electron-poor as well as
Jean-François Paquin et al.
Organic letters, 7(17), 3821-3824 (2005-08-12)
A general route to enantioenriched tert-butyl 3,3-diarylpropanoates is presented. These useful building blocks are prepared via an asymmetric rhodium-catalyzed conjugate addition of arylboronic acids to unsaturated tert-butyl esters in the presence of chiral dienes as ligands. The addition of both

Articles

The selective preparation of diarylmethine stereogenic centers is a challenging endeavor in chemical synthesis, especially when there is little differentiation between the two aryl groups

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service