649597
Poly(bis(2,2,2-trifluoroethoxy)phosphazene)
Synonym(s):
Poly[bis(trifluoroethoxy)phosphazene]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
solid
General description
Poly(bis(2,2,2-trifluoroethoxy)phosphazene), a highly hydrophobic fluorinated organic-soluble polymer is made up of trifluoroethoxy side groups that are connected to alternating phosphorus and nitrogen atoms. Polyphosphazenes are well suited for a variety of surface applications due to their properties such as hydrophobicity, resistance to many chemicals, bio inertness, high fire resistance, and radiation stability. Due to these excellent properties of poly[bis(2,2,2-trifluoroethoxy)phosphazene used in various fields such as membrane research, biomedical materials, battery electrolytes, surface coatings, elastomers, and nonlinear optical (NLO) materials.
Application
A solution of poly[bis(2,2,2-trifluoroethoxy)phosphazene] (PTFEP) in tetrahydrofuran, methylethyl ketone and acetone may be used to fabricate nanofibers of PTFEP. Electrospun PTFEP nanofiber mat may find uses in tissue engineering, drug delivery, filters and membranes.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biomaterials, 14(6), 430-436 (1993-05-01)
Investigations were carried out on the metathetical exchange reaction between the -O-CH2CF3 moieties of poly(bis(trifluoroethoxy)phosphazene) (PTFP), in the state of slightly swollen films, and the alkoxide ions derived from methoxypolyethylene glycol (MPEG) of molecular mass ranging from 350 to 5000
New amphiphilic poly [bis (2, 2, 2-trifluoroethoxy) phosphazene]/poly (propylene glycol) triblock copolymers: Synthesis and micellar characteristics
Macromolecules, 39(24), 8334-8338 (2006)
New second-order nonlinear optical polyphosphazenes: Convenient postfunctionalization synthetic approach and application of the concept of suitable isolation group
Dyes and Pigments, 84(3), 229-236 (2010)
Journal of vascular and interventional radiology : JVIR, 20(12), 1597-1607 (2009-12-01)
To determine the arterial distribution pattern of the embolic agent Embozene within the porcine kidney and compare it with those of other spherical embolic agents. Embozene, Embosphere, Bead Block, and Contour SE in size classes of 100-300 microm, 500-700 microm
Medical science monitor : international medical journal of experimental and clinical research, 14(2), BR35-BR40 (2008-01-30)
Clinical success of bone implants is critically related to the interaction between the implant surface and the surrounding tissue. The polymer poly(bis(trifluoroethoxy)phosphazene) (PTFEP) is a promising, highly biocompatible surface coating which also inhibits the adsorption of granulocytes, macrophages, inflammatory cells
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service