Skip to Content
Merck
All Photos(1)

Documents

578843

Sigma-Aldrich

3-(N-Boc-amino)phenylboronic acid pinacol ester

97%

Synonym(s):

3-(Boc-amino)phenylboronic acid pinacol ester, tert-Butyl-N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26BNO4
CAS Number:
Molecular Weight:
319.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

148-153 °C (lit.)

SMILES string

CC(C)(C)OC(=O)Nc1cccc(c1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C17H26BNO4/c1-15(2,3)21-14(20)19-13-10-8-9-12(11-13)18-22-16(4,5)17(6,7)23-18/h8-11H,1-7H3,(H,19,20)

InChI key

ANQAOGOIWVMGCH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service