539600
5-Amino-2-methoxybenzenesulfonic acid
97%
Synonym(s):
p-Anisidine-3-sulfonic acid
About This Item
Recommended Products
Assay
97%
mp
314-318 °C (lit.)
SMILES string
COc1ccc(N)cc1S(O)(=O)=O
InChI
1S/C7H9NO4S/c1-12-6-3-2-5(8)4-7(6)13(9,10)11/h2-4H,8H2,1H3,(H,9,10,11)
InChI key
JXZGTFLJFKLVAX-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service