Skip to Content
Merck
All Photos(1)

Key Documents

522961

Sigma-Aldrich

Dinonylnaphthalenesulfonic acid solution

50 wt. % in heptane

Synonym(s):

DNNSA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H19)2C10H5SO3H
CAS Number:
Molecular Weight:
460.71
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

concentration

50 wt. % in heptane

refractive index

n20/D 1.465

bp

94-99 °C

solubility

H2O: insoluble
alcohols, glycol ethers, glycols, esters, ketones, aromatic and aliphatic hydrocarbons: soluble

density

0.852 g/mL at 25 °C

SMILES string

CCCCCCCCCc1ccc2cc(CCCCCCCCC)cc(c2c1)S(O)(=O)=O

Looking for similar products? Visit Product Comparison Guide

Application

Dopant for conducting polymers hydrophobic acid catalyst for amino cross-linked coatings

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service