Skip to Content
Merck
All Photos(1)

Documents

453323

Sigma-Aldrich

Dimethyl sulfoxide-d6

"100%", 99.96 atom % D

Synonym(s):

(Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2SO
CAS Number:
Molecular Weight:
84.17
Beilstein:
1237248
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:

vapor pressure

0.42 mmHg ( 20 °C)

Quality Level

isotopic purity

99.96 atom % D

Assay

≥99.0%

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %

packaging

pk of 10 × 0.25 mL

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.476 (lit.)

bp

189 °C (lit.)

mp

20.2 °C (lit.)

density

1.190 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3/i1D3,2D3

InChI key

IAZDPXIOMUYVGZ-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyl sulfoxide-d6 (DMSO-d6) is a deuterated NMR solvent. It undergoes photodissociation to generate CD3 radical photoproducts, which have been analyzed by infrared diode laserabsorption spectroscopy. Dissociation dynamics of DMSO-d6 at 193nm was examined using photo fragment translational spectroscopy method.

Application

Dimethyl sulfoxide-d6 may be used as an NMR solvent for 1H and 13C NMR experiments.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Unraveling the dissociation of dimethyl sulfoxide following absorption at 193 nm.
Blank DA, et al.
J. Chem. Phys. , 106(2), 539-550 (1997)
Solution forms of an antitumor cyclic hexapeptide, RA-VII in dimethyl sulfoxide-d6 from nuclear magnetic resonance studies.
Itokawa H, et al.
Chemical & Pharmaceutical Bulletin, 40(4), 1050-1052 (1992)
Diode laser measurements of CD3 quantum yields and internal energy for the dissociation of dimethyl sulfoxide-d6.
Rudolph RN, et al.
J. Chem. Phys. , 106(4), 1346-1352 (1997)
Papawee Suabjakyong et al.
PloS one, 10(3), e0122733-e0122733 (2015-03-27)
The basidiomycetous mushroom Phellinus igniarius (L.) Quel. has been used as traditional medicine in various Asian countries for many years. Although many reports exist on its anti-oxidative and anti-inflammatory activities and therapeutic effects against various diseases, our current knowledge of
Barbara Parrino et al.
European journal of medicinal chemistry, 94, 367-377 (2015-03-18)
Three new ring systems, pyrido[2',3':3,4]pyrrolo[1,2-a]quinoxalines, pyrido[3',2':3,4]pyrrolo[1,2-a]quinoxalines and pyrido[2',3':5,6]pyrazino[2,1-a]isoindoles, were synthesized through an aza-substitution on the already active isoindolo-quinoxaline system and in particular in the position 7 or 4 of the isoindole moiety and in position 5 of the quinoxaline portion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service